Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 55 ESD Tips, 2 mm Tip width, 0.7 mm Tip thickness, 115 mm OAL
SDP
Catalog No. 501927511
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 55-ESD tips. Tips attach with screws. Length is 115 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 2 mm
Tip thickness: 0.7 mm
OAL: 115 mm
Catalog No. 50-192-7511 Supplier Electron Microscopy Sciences Supplier No. 10004E55SM
These tweezers have an anti-magnetic WA1 handle with replaceable 55-ESD tips. Tips attach with screws. Length is 115 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 2 mm
Tip thickness: 0.7 mm
OAL: 115 mm