Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 57 STD Tips, 1 mm Tip width, 0.7 mm Tip thickness, 120 mm OAL
SDP
Catalog No. 501927537
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 57-ESD tips. Tips attach with screws. Length is 120 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Grey STD
Tip width: 1 mm
Tip thickness: 0.7 mm
OAL: 120 mm
Catalog No. 50-192-7537 Supplier Electron Microscopy Sciences Supplier No. 10004S57SM
These tweezers have an anti-magnetic WA1 handle with replaceable 57-ESD tips. Tips attach with screws. Length is 120 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Grey STD
Tip width: 1 mm
Tip thickness: 0.7 mm
OAL: 120 mm