Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style F Buxus Tips, 2 mm Tip width, 0.2 mm Tip thickness, 130 mm OAL
SDP
Catalog No. 501927487
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceble F-BUIS tips made of boxwood (Buxus). Tips are attached with screws. Torque: cN.m 9 +/-1. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Boxwood (Buxus)
Tip width: 2 mm
Tip thickness: 0.2 mm
OAL: 130 mm
Catalog No. 50-192-7487 Supplier Electron Microscopy Sciences Supplier No. 10004BFSM
These tweezers have an anti-magnetic WA1 handle with replaceble F-BUIS tips made of boxwood (Buxus). Tips are attached with screws. Torque: cN.m 9 +/-1. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Boxwood (Buxus)
Tip width: 2 mm
Tip thickness: 0.2 mm
OAL: 130 mm