Learn More
Electron Microscopy Sciences Dumont WA2 Tweezer Handle with Style CA275 Ceramic Tips, 0.4 mm Tip width, 0.3 mm Tip thickness, 133 mm OAL
SDP
Catalog No. 501927539
Encompass_Preferred
These tweezers have an anti-magnetic WA2 handle with replaceable CA275-CW tips. Tips attach with screws. Length is 133 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Black Ceramic
Tip width: 0.4 mm
Tip thickness: 0.3 mm
OAL: 133 mm
Catalog No. 50-192-7539 Supplier Electron Microscopy Sciences Supplier No. 10004CA275SM
These tweezers have an anti-magnetic WA2 handle with replaceable CA275-CW tips. Tips attach with screws. Length is 133 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Black Ceramic
Tip width: 0.4 mm
Tip thickness: 0.3 mm
OAL: 133 mm