Learn More
MSC Buyers Products Spinner, 9 in, Polyurethane, TGSUV1A/TGSUV1B Compatible
Catalog No. 502836867
Encompass
This replacement spinner is designed for use with SaltDogg TGSUV1A and TGSUV1B spreaders. Made from durable polyurethane, it features a 9 in diameter, ensuring reliable performance in various spreading applications.
  • Designed for SaltDogg TGSUV1A and TGSUV1B spreaders.
  • Constructed from durable polyurethane.
  • Features a 9 in diameter.
  • Ideal as a replacement part.
Catalog No. 50-283-6867 Supplier MSC Supplier No. 56033897
This replacement spinner is designed for use with SaltDogg TGSUV1A and TGSUV1B spreaders. Made from durable polyurethane, it features a 9 in diameter, ensuring reliable performance in various spreading applications.
  • Designed for SaltDogg TGSUV1A and TGSUV1B spreaders.
  • Constructed from durable polyurethane.
  • Features a 9 in diameter.
  • Ideal as a replacement part.