Learn More
Chemglass Life Sciences Replacement PTFE Auger B
SDP
Catalog No. CG172322
Encompass_Preferred
Replacement PTFE auger B for CG1723 series of addition funnels. Compatible with the 250mL and 500mL size powder addition funnels (CG-1723-13 to CG-1723-17). NOT COMPATIBLE WITH SMALLER FUNNELS. For smaller powder addition funnel's replacement auger, see Auger A replacement (CG-1723-20).
Catalog No. CG172322 Supplier Chemglass Life Sciences Supplier No. CG172322
Replacement PTFE auger B for CG1723 series of addition funnels. Compatible with the 250mL and 500mL size powder addition funnels (CG-1723-13 to CG-1723-17). NOT COMPATIBLE WITH SMALLER FUNNELS. For smaller powder addition funnel's replacement auger, see Auger A replacement (CG-1723-20).