Learn More
Chemglass Life Sciences Replacement Viton O-Rings for 3/8" Unions, Airfree, Schlenk
SDP
Catalog No. AF006721
Encompass_Preferred
Replacement Viton O-Rings for 3/8" Unions, Airfree, Schlenk. Replacement Viton O-Rings for item AF006705. O-Rings are 70 durometer Viton. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel.
Catalog No. AF006721 Supplier Chemglass Life Sciences Supplier No. AF006721
Replacement Viton O-Rings for 3/8" Unions, Airfree, Schlenk. Replacement Viton O-Rings for item AF006705. O-Rings are 70 durometer Viton. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel.