Learn More
Meso Scale Discovery R-PLEX Human MIP-4 Assay, singleplex format, 5 plate size, 2.2 pg/mL lower limit of detection, 96-well plate, Human Macrophage inflammatory protein 4
SDP
Catalog No. NC1996604
Encompass
R-PLEX Human MIP-4 Assay, singleplex format, 5 plate size, 2.2 pg/mL lower limit of detection, 96-well plate, Human Macrophage inflammatory protein 4
Catalog No. NC1996604 Supplier Meso Scale Discovery Supplier No. K1512HR2
R-PLEX Human MIP-4 Assay, singleplex format, 5 plate size, 2.2 pg/mL lower limit of detection, 96-well plate, Human Macrophage inflammatory protein 4