Learn More
Meso Scale Discovery RPLEX Hu ADPrib cyclase ASY
SDP
Catalog No. 5000081971
Encompass
The R-Plex Human ADP-ribosyl Cyclase Assay from Meso Scale Discovery is a 96-well plate assay measuring ADP-ribosyl cyclase activity in human samples It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range This quantitative immunoassay detects cyclic ADP-ribose (cADPR) and requires 50 L of sample per well The kit includes a 96-well carbon electrode plate detection antibody labeled with MSD SULFO-TAG read buffer wash buffer blocking solution calibration standard and positive control
Catalog No. 50-000-81971 Supplier Meso Scale Discovery Supplier No. K151ALTR2
Add to Cart
Add to Cart
The R-Plex Human ADP-ribosyl Cyclase Assay from Meso Scale Discovery is a 96-well plate assay measuring ADP-ribosyl cyclase activity in human samples It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range This quantitative immunoassay detects cyclic ADP-ribose (cADPR) and requires 50 L of sample per well The kit includes a 96-well carbon electrode plate detection antibody labeled with MSD SULFO-TAG read buffer wash buffer blocking solution calibration standard and positive control