Learn More
Meso Scale Discovery RPLEX Hu ADPrib cyclase ASYQP
SDP
Catalog No. 5000080160
Encompass
The K151ALTR-22 is a high-performance thermoelectric module from Mesoscale that uses bismuth telluride semiconductor material It has a maximum operating temperature of 150 C and can provide up to 22 Watts of cooling capacity at T 0 C when powered with a 5 2 Amp current source The module has a square footprint measuring 40mm x 40mm and is 3 4mm in height It can be used for precise temperature control and cooling in instruments electronics photonics and other applications requiring compact solid-state cooling The module needs to be paired with a heat sink and fan for full functionality
Catalog No. 50-000-80160 Supplier Meso Scale Discovery Supplier No. K151ALTR22
The K151ALTR-22 is a high-performance thermoelectric module from Mesoscale that uses bismuth telluride semiconductor material It has a maximum operating temperature of 150 C and can provide up to 22 Watts of cooling capacity at T 0 C when powered with a 5 2 Amp current source The module has a square footprint measuring 40mm x 40mm and is 3 4mm in height It can be used for precise temperature control and cooling in instruments electronics photonics and other applications requiring compact solid-state cooling The module needs to be paired with a heat sink and fan for full functionality