Learn More
Meso Scale Discovery RPLEX Hu Antileukoprotei ASYQP
SDP
Catalog No. 5000079654
Encompass
The R-PLEX Human Antileukoproteinase Assay Kit from Meso Scale Diagnostics measures human antileukoproteinase (ALP) in various biological fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent It has a sensitivity of 19 pg/mL and an upper limit of 10 000 pg/mL detecting native and recombinant ALP in 3 5 hours
Catalog No. 50-000-79654 Supplier Meso Scale Discovery Supplier No. K151G4R22
The R-PLEX Human Antileukoproteinase Assay Kit from Meso Scale Diagnostics measures human antileukoproteinase (ALP) in various biological fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent It has a sensitivity of 19 pg/mL and an upper limit of 10 000 pg/mL detecting native and recombinant ALP in 3 5 hours