Learn More
Meso Scale Discovery RPLEX Hu Antileukoproteina ASY
SDP
Catalog No. 5000081611
Encompass
The R-PLEX Human Antileukoproteinase Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of human antileukoproteinase (ALP) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of human ALP This assay can be used to study ALP levels in disease research biomarker discovery and drug development With high sensitivity broad dynamic range and minimal matrix interference the R-PLEX ALP assay enables robust and reproducible quantification of this important serine protease inhibitor
Catalog No. 50-000-81611 Supplier Meso Scale Discovery Supplier No. K151G4R2
The R-PLEX Human Antileukoproteinase Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of human antileukoproteinase (ALP) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of human ALP This assay can be used to study ALP levels in disease research biomarker discovery and drug development With high sensitivity broad dynamic range and minimal matrix interference the R-PLEX ALP assay enables robust and reproducible quantification of this important serine protease inhibitor