Learn More
Meso Scale Discovery RPLEX Hu Cathepsin S TTL ASY
SDP
Catalog No. 5000078466
Encompass
The R-PLEX Hu Cathepsin S Total Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Cathepsin S in cell and tissue lysates This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and Calibrator This assay allows for the simple and rapid measurement of Cathepsin S across multiple samples
Catalog No. 50-000-78466 Supplier Meso Scale Discovery Supplier No. K151ALXR2
The R-PLEX Hu Cathepsin S Total Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Cathepsin S in cell and tissue lysates This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and Calibrator This assay allows for the simple and rapid measurement of Cathepsin S across multiple samples