Learn More
Meso Scale Discovery RPLEX Hu CathepsinB TTL ASYQP
SDP
Catalog No. 5000079527
Encompass
The R-PLEX Hu Cathepsin B Total Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of total human Cathepsin B in cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Diagnostics SECTOR Imagers The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent wash buffer and lyophilized calibrator
Catalog No. 50-000-79527 Supplier Meso Scale Discovery Supplier No. K151ACAR22
The R-PLEX Hu Cathepsin B Total Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of total human Cathepsin B in cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Diagnostics SECTOR Imagers The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent wash buffer and lyophilized calibrator