Learn More
Meso Scale Discovery RPLEX Hu PAF acetylhydro ASYQP
SDP
Catalog No. 5000079024
Encompass
The R-PLEX Hu PAF Acetyl Hydrolase Assay is a 96-well plate kit for measuring PAF acetylhydrolase activity in human serum plasma and cell lysates It includes all necessary reagents like substrate buffer and standards The assay uses hydrolysis of 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine by PAF acetylhydrolase producing choline detected via a coupled enzymatic reaction with choline oxidase It supports high-throughput screening with a detection limit of 0 4 mU/mL and takes about 3 hours This kit is sensitive and convenient for studying PAF acetylhydrolase in human samples
Catalog No. 50-000-79024 Supplier Meso Scale Discovery Supplier No. K151ABXR22
The R-PLEX Hu PAF Acetyl Hydrolase Assay is a 96-well plate kit for measuring PAF acetylhydrolase activity in human serum plasma and cell lysates It includes all necessary reagents like substrate buffer and standards The assay uses hydrolysis of 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine by PAF acetylhydrolase producing choline detected via a coupled enzymatic reaction with choline oxidase It supports high-throughput screening with a detection limit of 0 4 mU/mL and takes about 3 hours This kit is sensitive and convenient for studying PAF acetylhydrolase in human samples