Learn More
Meso Scale Discovery RPLEX Hu PAF acetylhydrola ASY
SDP
Catalog No. 5000078223
Encompass
The R-PLEX Hu PAF Acetyl Hydrolase Assay is a 96-well plate assay kit for measuring PAF acetylhydrolase activity in human serum plasma and cell lysates The kit contains a PAF substrate that is hydrolyzed by PAF-AH generating free thiol groups that can be detected using a maleimide dye The kit includes all necessary reagents and can process up to 39 samples in duplicate The assay is rapid sensitive and reproducible It is useful for studying PAF metabolism and the role of PAF-AH in inflammation and cardiovascular disease
Catalog No. 50-000-78223 Supplier Meso Scale Discovery Supplier No. K151ABXR2
The R-PLEX Hu PAF Acetyl Hydrolase Assay is a 96-well plate assay kit for measuring PAF acetylhydrolase activity in human serum plasma and cell lysates The kit contains a PAF substrate that is hydrolyzed by PAF-AH generating free thiol groups that can be detected using a maleimide dye The kit includes all necessary reagents and can process up to 39 samples in duplicate The assay is rapid sensitive and reproducible It is useful for studying PAF metabolism and the role of PAF-AH in inflammation and cardiovascular disease