Learn More
Meso Scale Discovery RPLEX Hu proCollagen alphaASY
SDP
Catalog No. 5000079526
Encompass
The R-PLEX Hu Procollagen Alpha 1 Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of procollagen alpha 1 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR technology is sensitive rapid and requires only 50 L sample volume The assay has a typical lower limit of quantitation of 0 244 ng/mL and an upper limit of quantitation of 200 ng/mL It includes a 96-well plate coated with capture antibody detection antibody Read Buffer and Calibrator This quantitative immunoassay provides a tool for studying collagen synthesis and turnover
Catalog No. 50-000-79526 Supplier Meso Scale Discovery Supplier No. K151ABYR22
The R-PLEX Hu Procollagen Alpha 1 Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of procollagen alpha 1 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR technology is sensitive rapid and requires only 50 L sample volume The assay has a typical lower limit of quantitation of 0 244 ng/mL and an upper limit of quantitation of 200 ng/mL It includes a 96-well plate coated with capture antibody detection antibody Read Buffer and Calibrator This quantitative immunoassay provides a tool for studying collagen synthesis and turnover