Learn More
Meso Scale Discovery RPLEX Hu Troponin I crdc ASYQP
SDP
Catalog No. 5000078391
Encompass
The R-Plex Human Troponin I Cardiac Assay from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures cardiac troponin I in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume This assay allows for the rapid and efficient analysis of cardiac troponin I a biomarker used to aid in the diagnosis of acute myocardial infarction With a measuring range of 0 5-2500 pg/mL and an LLOQ of 1 1 pg/mL this assay kit enables the sensitive and accurate quantification of cardiac troponin I levels
Catalog No. 50-000-78391 Supplier Meso Scale Discovery Supplier No. K1513LR22
The R-Plex Human Troponin I Cardiac Assay from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures cardiac troponin I in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume This assay allows for the rapid and efficient analysis of cardiac troponin I a biomarker used to aid in the diagnosis of acute myocardial infarction With a measuring range of 0 5-2500 pg/mL and an LLOQ of 1 1 pg/mL this assay kit enables the sensitive and accurate quantification of cardiac troponin I levels