Learn More
Meso Scale Discovery RPLEX HUM 41BBL/TNFSF9 ASYQP
SDP
Catalog No. 5000082094
Encompass
The R-PLEX Human 4-1BBL (TNFSF9) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human 4-1BBL in serum plasma and cell culture supernatants It utilizes electrochemiluminescence for high sensitivity and broad dynamic range This assay allows for the quantification of natural and recombinant human 4-1BBL with a lower limit of detection of 1 15 pg/mL It provides reagents for 1 plate which can test up to 80 samples in duplicate
Catalog No. 50-000-82094 Supplier Meso Scale Discovery Supplier No. K151ALLR22
The R-PLEX Human 4-1BBL (TNFSF9) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human 4-1BBL in serum plasma and cell culture supernatants It utilizes electrochemiluminescence for high sensitivity and broad dynamic range This assay allows for the quantification of natural and recombinant human 4-1BBL with a lower limit of detection of 1 15 pg/mL It provides reagents for 1 plate which can test up to 80 samples in duplicate