Learn More
Meso Scale Discovery RPLEX HUM 6CKine/CCL21 ASYQP
SDP
Catalog No. 5000079293
Encompass
The R-PLEX Human CCL21 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CCL21 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantitation of CCL21 across a wide range of concentrations
Catalog No. 50-000-79293 Supplier Meso Scale Discovery Supplier No. K151M1R22
The R-PLEX Human CCL21 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CCL21 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantitation of CCL21 across a wide range of concentrations