Learn More
Meso Scale Discovery RPLEX HUM Alphaamylase 1 ASY
SDP
Catalog No. 5000078386
Encompass
The R-PLEX Human Alpha-Amylase 1 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human alpha-amylase 1 in serum plasma saliva and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluent read buffer and wash buffer Sensitivity is 0 08 ng/mL with an upper quantification limit of 1500 ng/mL
Catalog No. 50-000-78386 Supplier Meso Scale Discovery Supplier No. K1513CR2
The R-PLEX Human Alpha-Amylase 1 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human alpha-amylase 1 in serum plasma saliva and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluent read buffer and wash buffer Sensitivity is 0 08 ng/mL with an upper quantification limit of 1500 ng/mL