Learn More
Meso Scale Discovery RPLEX HUM Angiopoietin2 ASYQP
SDP
Catalog No. 5000078786
Encompass
The R-PLEX Human Angiopoietin-2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-2 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay Results are obtained in as little as 3 5 hours This assay is for research use only
Catalog No. 50-000-78786 Supplier Meso Scale Discovery Supplier No. K151YRR22
The R-PLEX Human Angiopoietin-2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-2 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay Results are obtained in as little as 3 5 hours This assay is for research use only