Learn More
Meso Scale Discovery RPLEX HUM BclX/Bcl2L1 ASY
SDP
Catalog No. 5000078887
Encompass
The R-PLEX Human Bcl-xL/Bcl2-L-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-xL and Bcl2-L-1 protein levels in human samples such as serum plasma and cell lysates The assay utilizes electrochemiluminescence (ECL) detection and provides high sensitivity broad dynamic range and rapid results This multiplex assay allows you to measure both proteins in a single well saving sample volume and time The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay Results are obtained in as little as 3 5 hours
Catalog No. 50-000-78887 Supplier Meso Scale Discovery Supplier No. K1516ZR2
The R-PLEX Human Bcl-xL/Bcl2-L-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-xL and Bcl2-L-1 protein levels in human samples such as serum plasma and cell lysates The assay utilizes electrochemiluminescence (ECL) detection and provides high sensitivity broad dynamic range and rapid results This multiplex assay allows you to measure both proteins in a single well saving sample volume and time The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay Results are obtained in as little as 3 5 hours