Learn More
Meso Scale Discovery RPLEX HUM BclX/Bcl2L1 ASYQP
SDP
Catalog No. 5000078888
Encompass
The R-PLEX Human Bcl-xL/Bcl2-L-1 Assay Kit from Meso Scale Discovery measures Bcl-xL and Bcl2-L-1 protein levels in human samples like serum plasma and cell lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results analyzed on MSD systems offer insights into apoptotic regulation pathways with high sensitivity and minimal sample needs
Catalog No. 50-000-78888 Supplier Meso Scale Discovery Supplier No. K1516ZR22
The R-PLEX Human Bcl-xL/Bcl2-L-1 Assay Kit from Meso Scale Discovery measures Bcl-xL and Bcl2-L-1 protein levels in human samples like serum plasma and cell lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results analyzed on MSD systems offer insights into apoptotic regulation pathways with high sensitivity and minimal sample needs