Learn More
Meso Scale Discovery RPLEX HUM C1q receptor ASYQP
SDP
Catalog No. 5000080391
Encompass
The R-PLEX Human C1q Receptor Assay Kit from Meso Scale Discovery is used to quantitatively measure C1q receptor levels in human serum plasma cell culture supernatants and other biological samples This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The C1q receptor plays an important role in the clearance of apoptotic cells and immune complexes Monitoring levels of this receptor may have research and diagnostic utility
Catalog No. 50-000-80391 Supplier Meso Scale Discovery Supplier No. K151AAHR22
The R-PLEX Human C1q Receptor Assay Kit from Meso Scale Discovery is used to quantitatively measure C1q receptor levels in human serum plasma cell culture supernatants and other biological samples This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The C1q receptor plays an important role in the clearance of apoptotic cells and immune complexes Monitoring levels of this receptor may have research and diagnostic utility