Learn More
Meso Scale Discovery RPLEX HUM Calprotectin ASYQP
SDP
Catalog No. 5000081663
Encompass
The R-PLEX Human Calprotectin Assay by Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring calprotectin in human serum plasma and other fluids It uses the SECTOR Imager 6000 for high sensitivity and a broad dynamic range enabling rapid and accurate quantification of calprotectin a biomarker for inflammation The assay is highly specific requires a small sample and delivers results in hours The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for analysis
Catalog No. 50-000-81663 Supplier Meso Scale Discovery Supplier No. K151AJYR22
The R-PLEX Human Calprotectin Assay by Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring calprotectin in human serum plasma and other fluids It uses the SECTOR Imager 6000 for high sensitivity and a broad dynamic range enabling rapid and accurate quantification of calprotectin a biomarker for inflammation The assay is highly specific requires a small sample and delivers results in hours The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for analysis