Learn More
Meso Scale Discovery RPLEX HUM Cathepsin L2 ASYQP
SDP
Catalog No. 5000081618
Encompass
The R-PLEX Human Cathepsin L2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Cathepsin L2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay It can detect Cathepsin L2 concentrations as low as 3 2 pg/mL with an upper limit of 100 000 pg/mL
Catalog No. 50-000-81618 Supplier Meso Scale Discovery Supplier No. K151AJGR22
The R-PLEX Human Cathepsin L2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Cathepsin L2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay It can detect Cathepsin L2 concentrations as low as 3 2 pg/mL with an upper limit of 100 000 pg/mL