Learn More
Meso Scale Discovery RPLEX HUM ChromograninA ASY
SDP
Catalog No. 5000082471
Encompass
The Meso Scale Discovery R-PLEX Human Chromogranin A Assay is a multiplex immunoassay panel that measures Chromogranin A levels in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of Chromogranin A which can aid in the diagnosis of neuroendocrine tumors heart failure and other conditions involving the regulated secretory pathway With minimal hands-on time required this assay enables rapid and reliable quantification of Chromogranin A levels
Catalog No. 50-000-82471 Supplier Meso Scale Discovery Supplier No. K151C5R2
The Meso Scale Discovery R-PLEX Human Chromogranin A Assay is a multiplex immunoassay panel that measures Chromogranin A levels in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of Chromogranin A which can aid in the diagnosis of neuroendocrine tumors heart failure and other conditions involving the regulated secretory pathway With minimal hands-on time required this assay enables rapid and reliable quantification of Chromogranin A levels