Learn More
Meso Scale Discovery RPLEX HUM ChromograninA ASYQP
SDP
Catalog No. 5000079693
Encompass
The Meso Scale Discovery R-PLEX Human Chromogranin A Assay is a multiplex immunoassay that measures Chromogranin A in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L of sample it runs on MSD QuickPlex SQ 120 or standard MSD instruments The detection range is 0 08 to 2500 ng/mL This assay quantifies Chromogranin A useful as a biomarker for neuroendocrine tumors and other diseases
Catalog No. 50-000-79693 Supplier Meso Scale Discovery Supplier No. K151C5R22
The Meso Scale Discovery R-PLEX Human Chromogranin A Assay is a multiplex immunoassay that measures Chromogranin A in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L of sample it runs on MSD QuickPlex SQ 120 or standard MSD instruments The detection range is 0 08 to 2500 ng/mL This assay quantifies Chromogranin A useful as a biomarker for neuroendocrine tumors and other diseases