Learn More
Meso Scale Discovery RPLEX HUM CK Beta 81/CCL23 ASY
SDP
Catalog No. 5000078741
Encompass
The Human CK-beta-8-1/CCL23 DuoSet ELISA is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CK-beta-8-1/CCL23 in cell culture supernatants serum plasma and other biological fluids This immunoassay contains recombinant human CK-beta-8-1/CCL23 and antibodies raised against the recombinant factor allowing for the detection of natural and recombinant human CK-beta-8-1/CCL23 The ELISA has a sensitivity of 7 8 pg/mL and a detection range of 15 6-1000 pg/mL This product can be used to measure CK-beta-8-1/CCL23 levels for the study of chemokine biology and inflammation
Catalog No. 50-000-78741 Supplier Meso Scale Discovery Supplier No. K151C1R2
The Human CK-beta-8-1/CCL23 DuoSet ELISA is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CK-beta-8-1/CCL23 in cell culture supernatants serum plasma and other biological fluids This immunoassay contains recombinant human CK-beta-8-1/CCL23 and antibodies raised against the recombinant factor allowing for the detection of natural and recombinant human CK-beta-8-1/CCL23 The ELISA has a sensitivity of 7 8 pg/mL and a detection range of 15 6-1000 pg/mL This product can be used to measure CK-beta-8-1/CCL23 levels for the study of chemokine biology and inflammation