Learn More
Meso Scale Discovery RPLEX HUM Complement C3 ASYQP
SDP
Catalog No. 5000081804
Encompass
The R-PLEX Human Complement C3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures levels of complement component C3 in human serum plasma and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator This assay allows for the quantitative measurement of complement C3 levels which can aid in the diagnosis of immunological and inflammatory conditions
Catalog No. 50-000-81804 Supplier Meso Scale Discovery Supplier No. K151XYR22
The R-PLEX Human Complement C3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures levels of complement component C3 in human serum plasma and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator This assay allows for the quantitative measurement of complement C3 levels which can aid in the diagnosis of immunological and inflammatory conditions