Learn More
Meso Scale Discovery RPLEX HUM Complement C3a ASY
SDP
Catalog No. 5000079717
Encompass
The R-PLEX Human Complement C3a Assay from Meso Scale Discovery is a multiplex immunoassay that measures C3a in human serum plasma and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 1 3 pg/mL This product enables the quantification of complement activation through measurement of C3a It is suitable for research use only
Catalog No. 50-000-79717 Supplier Meso Scale Discovery Supplier No. K151V0R2
The R-PLEX Human Complement C3a Assay from Meso Scale Discovery is a multiplex immunoassay that measures C3a in human serum plasma and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 1 3 pg/mL This product enables the quantification of complement activation through measurement of C3a It is suitable for research use only