Learn More
Meso Scale Discovery RPLEX HUM Complement C3a ASYQP
SDP
Catalog No. 5000079122
Encompass
The R-PLEX Human Complement C3a Assay from Meso Scale Discovery is a multiplex immunoassay that measures C3a in human serum plasma and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The C3a assay measures complement activation and can be used to monitor inflammation and autoimmune diseases This quantitative assay has high reproducibility and requires 2 5 hours to complete
Catalog No. 50-000-79122 Supplier Meso Scale Discovery Supplier No. K151V0R22
The R-PLEX Human Complement C3a Assay from Meso Scale Discovery is a multiplex immunoassay that measures C3a in human serum plasma and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The C3a assay measures complement activation and can be used to monitor inflammation and autoimmune diseases This quantitative assay has high reproducibility and requires 2 5 hours to complete