Learn More
Meso Scale Discovery RPLEX HUM Complement C5a ASY
SDP
Catalog No. 5000080796
Encompass
The R-PLEX Human Complement C5a Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human complement component C5a in serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity broad dynamic range and rapid results This research-use-only kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure C5a concentrations between 0 3-2500 pg/mL with an assay time of just 3 5 hours
Catalog No. 50-000-80796 Supplier Meso Scale Discovery Supplier No. K151K4R2
The R-PLEX Human Complement C5a Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human complement component C5a in serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity broad dynamic range and rapid results This research-use-only kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure C5a concentrations between 0 3-2500 pg/mL with an assay time of just 3 5 hours