Learn More
Meso Scale Discovery RPLEX HUM Cytokeratin19 ASY
SDP
Catalog No. 5000079426
Encompass
The R-PLEX Human Cytokeratin 19 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures cytokeratin 19 levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay It can be used for cancer biomarker research and measuring treatment response The assay has a lower limit of detection of 0 016 ng/mL and an upper limit of quantification of 400 ng/mL for cytokeratin 19
Catalog No. 50-000-79426 Supplier Meso Scale Discovery Supplier No. K151A9R2
The R-PLEX Human Cytokeratin 19 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures cytokeratin 19 levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay It can be used for cancer biomarker research and measuring treatment response The assay has a lower limit of detection of 0 016 ng/mL and an upper limit of quantification of 400 ng/mL for cytokeratin 19