Learn More
Meso Scale Discovery RPLEX HUM Cytokeratin19 ASYQP
SDP
Catalog No. 5000078958
Encompass
The R-PLEX Human Cytokeratin 19 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure Cytokeratin 19 levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time to results This assay kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator It can detect Cytokeratin 19 levels as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL The total assay time is 2 5 hours
Catalog No. 50-000-78958 Supplier Meso Scale Discovery Supplier No. K151A9R22
The R-PLEX Human Cytokeratin 19 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure Cytokeratin 19 levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time to results This assay kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator It can detect Cytokeratin 19 levels as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL The total assay time is 2 5 hours