Learn More
Meso Scale Discovery RPLEX HUM Cytokeratin8 ASYQP
SDP
Catalog No. 5000078821
Encompass
The R-PLEX Human Cytokeratin 8 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure cytokeratin 8 levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent immunoassay method based on MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent It can detect cytokeratin 8 levels as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL
Catalog No. 50-000-78821 Supplier Meso Scale Discovery Supplier No. K1510CR22
The R-PLEX Human Cytokeratin 8 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure cytokeratin 8 levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent immunoassay method based on MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent It can detect cytokeratin 8 levels as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL