Learn More
Meso Scale Discovery RPLEX HUM Endothelin1 ASYQP
SDP
Catalog No. 5000081953
Encompass
The R-PLEX Human Endothelin-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures Endothelin-1 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies a detection antibody solution read buffer calibrator diluent and vials of calibrator The assay can detect Endothelin-1 in the range of 0 05-1000 pg/mL and requires only 25 L of sample
Catalog No. 50-000-81953 Supplier Meso Scale Discovery Supplier No. K151D4R22
The R-PLEX Human Endothelin-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures Endothelin-1 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies a detection antibody solution read buffer calibrator diluent and vials of calibrator The assay can detect Endothelin-1 in the range of 0 05-1000 pg/mL and requires only 25 L of sample