Learn More
Meso Scale Discovery RPLEX HUM ERK1/2 total ASYQP
SDP
Catalog No. 5000080149
Encompass
The R-PLEX Human ERK1/2 Total Assay Kit from Meso Scale Discovery is used to quantitatively measure total ERK1/2 levels in cell lysates The assay uses an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control lysate The assay can detect total ERK1/2 levels between 50 pg/mL to 50 000 pg/mL It is compatible with lysates from adherent and suspension cultured cells The kit provides all necessary reagents for the immunoassay
Catalog No. 50-000-80149 Supplier Meso Scale Discovery Supplier No. K151AKNR22
The R-PLEX Human ERK1/2 Total Assay Kit from Meso Scale Discovery is used to quantitatively measure total ERK1/2 levels in cell lysates The assay uses an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control lysate The assay can detect total ERK1/2 levels between 50 pg/mL to 50 000 pg/mL It is compatible with lysates from adherent and suspension cultured cells The kit provides all necessary reagents for the immunoassay