Learn More
Meso Scale Discovery RPLEX HUM FABP3/HFABP ASYQP
SDP
Catalog No. 5000078848
Encompass
The R-PLEX Human FABP3 (H-FABP) Assay Kit from Meso Scale Diagnostics is used to quantitatively measure human heart-type fatty acid binding protein (H-FABP) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range This multiplex kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrator diluent and lyophilized calibrator The assay takes 3 5 hours to complete
Catalog No. 50-000-78848 Supplier Meso Scale Discovery Supplier No. K1514TR22
The R-PLEX Human FABP3 (H-FABP) Assay Kit from Meso Scale Diagnostics is used to quantitatively measure human heart-type fatty acid binding protein (H-FABP) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range This multiplex kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrator diluent and lyophilized calibrator The assay takes 3 5 hours to complete