Learn More
Meso Scale Discovery RPLEX HUM FABP4/AFABP ASYQP
SDP
Catalog No. 5000078857
Encompass
The R-PLEX Human FABP4 (A-FABP) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Fatty Acid Binding Protein 4 (FABP4) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the rapid quantitation of FABP4 an important biomarker for obesity insulin resistance type 2 diabetes and cardiovascular disease With minimal hands-on time this kit enables the study of FABP4 biology and pathology
Catalog No. 50-000-78857 Supplier Meso Scale Discovery Supplier No. K151ABMR22
The R-PLEX Human FABP4 (A-FABP) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Fatty Acid Binding Protein 4 (FABP4) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the rapid quantitation of FABP4 an important biomarker for obesity insulin resistance type 2 diabetes and cardiovascular disease With minimal hands-on time this kit enables the study of FABP4 biology and pathology