Learn More
Meso Scale Discovery RPLEX HUM Fas soluble ASYQP
SDP
Catalog No. 5000081806
Encompass
The R-PLEX Human FAS Soluble Assay from Meso Scale Discovery is a multiplex immunoassay kit for quantifying fatty acid synthase (FAS) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescence detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies for human FAS detection antibodies calibrators and necessary reagents Measuring FAS levels aids in understanding metabolic diseases and cancer metabolism with results in 3 5 hours
Catalog No. 50-000-81806 Supplier Meso Scale Discovery Supplier No. K151ZPR22
The R-PLEX Human FAS Soluble Assay from Meso Scale Discovery is a multiplex immunoassay kit for quantifying fatty acid synthase (FAS) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescence detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies for human FAS detection antibodies calibrators and necessary reagents Measuring FAS levels aids in understanding metabolic diseases and cancer metabolism with results in 3 5 hours