Learn More
Meso Scale Discovery RPLEX HUM Fcepsilon RII ASY
SDP
Catalog No. 5000080384
Encompass
The R-PLEX Human FC Epsilon RII Assay Kit from Meso Scale Discovery measures free human IgE in samples like serum and plasma This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results similar to ELISA The kit includes a 96-well plate with anti-human IgE capture antibody SULFO-TAG labeled detection antibody assay diluent read buffer wash buffer and calibrator Sensitivity is 0 24 ng/mL with an upper quantitation limit of 1500 ng/mL IgE
Catalog No. 50-000-80384 Supplier Meso Scale Discovery Supplier No. K151AAQR2
The R-PLEX Human FC Epsilon RII Assay Kit from Meso Scale Discovery measures free human IgE in samples like serum and plasma This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results similar to ELISA The kit includes a 96-well plate with anti-human IgE capture antibody SULFO-TAG labeled detection antibody assay diluent read buffer wash buffer and calibrator Sensitivity is 0 24 ng/mL with an upper quantitation limit of 1500 ng/mL IgE