Learn More
Meso Scale Discovery RPLEX HUM Fcepsilon RII ASYQP
SDP
Catalog No. 5000079589
Encompass
The Meso Scale Discovery FC-Epsilon-RII Assay Kit is a highly sensitive immunoassay that quantitatively measures the soluble form of the Epsilon RII receptor in biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides a rapid 3 5 hour protocol from sample to results The FC-Epsilon-RII kit includes a 96-well plate coated with a mouse anti-human FC-Epsilon-RII capture antibody a ruthenylated mouse anti-human FC-Epsilon-RII detection antibody calibrators and an MSD SECTOR Imager for readout The assay has a sensitivity of 19 pg/mL and a broad dynamic range up to 10 000 pg/mL
Catalog No. 50-000-79589 Supplier Meso Scale Discovery Supplier No. K151AAQR22
The Meso Scale Discovery FC-Epsilon-RII Assay Kit is a highly sensitive immunoassay that quantitatively measures the soluble form of the Epsilon RII receptor in biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides a rapid 3 5 hour protocol from sample to results The FC-Epsilon-RII kit includes a 96-well plate coated with a mouse anti-human FC-Epsilon-RII capture antibody a ruthenylated mouse anti-human FC-Epsilon-RII detection antibody calibrators and an MSD SECTOR Imager for readout The assay has a sensitivity of 19 pg/mL and a broad dynamic range up to 10 000 pg/mL