Learn More
Meso Scale Discovery RPLEX HUM FGF acidic ASYQP
SDP
Catalog No. 5000079636
Encompass
The FGF Acidic ELISA Kit from Meso Scale Discovery is a sandwich immunoassay for the quantitative measurement of FGF Acidic in cell culture supernatants serum plasma and other biological fluids The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity wide dynamic range and rapid results The assay can detect FGF Acidic concentrations as low as 15 pg/mL with an upper limit of 50 000 pg/mL The total assay time is just under 3 5 hours This product could be useful for studying FGF Acidic signaling and disease research
Catalog No. 50-000-79636 Supplier Meso Scale Discovery Supplier No. K151W0R22
The FGF Acidic ELISA Kit from Meso Scale Discovery is a sandwich immunoassay for the quantitative measurement of FGF Acidic in cell culture supernatants serum plasma and other biological fluids The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity wide dynamic range and rapid results The assay can detect FGF Acidic concentrations as low as 15 pg/mL with an upper limit of 50 000 pg/mL The total assay time is just under 3 5 hours This product could be useful for studying FGF Acidic signaling and disease research