Learn More
Meso Scale Discovery RPLEX HUM Folate R alpha ASY
SDP
Catalog No. 5000079511
Encompass
The Meso Scale Discovery R-Plex Human Folate R-alpha Assay Kit is a multiplex immunoassay for measuring folate receptor alpha in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection for high sensitivity broad dynamic range and precision with low sample use The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and controls for quantifying folate receptor alpha a key biomarker in human samples
Catalog No. 50-000-79511 Supplier Meso Scale Discovery Supplier No. K151AASR2
The Meso Scale Discovery R-Plex Human Folate R-alpha Assay Kit is a multiplex immunoassay for measuring folate receptor alpha in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection for high sensitivity broad dynamic range and precision with low sample use The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and controls for quantifying folate receptor alpha a key biomarker in human samples