Learn More
Meso Scale Discovery RPLEX HUM Folate R alpha ASYQP
SDP
Catalog No. 5000079512
Encompass
The R-Plex Human Folate R-alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures folate receptor alpha in human serum plasma and cell culture supernatant samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay allows for the quantitative measurement of folate receptor alpha levels with a run time of approximately 3 5 hours
Catalog No. 50-000-79512 Supplier Meso Scale Discovery Supplier No. K151AASR22
The R-Plex Human Folate R-alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures folate receptor alpha in human serum plasma and cell culture supernatant samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay allows for the quantitative measurement of folate receptor alpha levels with a run time of approximately 3 5 hours