Learn More
Meso Scale Discovery RPLEX HUM Growth Hormone ASYQP
SDP
Catalog No. 5000079032
Encompass
The R-PLEX Human Growth Hormone Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures human growth hormone (hGH) in serum plasma cell culture supernatants and other biological sample types This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate pre-coated with capture antibody detection antibody calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay kit offers a simple rapid and robust method to measure hGH levels
Catalog No. 50-000-79032 Supplier Meso Scale Discovery Supplier No. K1519UR22
The R-PLEX Human Growth Hormone Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures human growth hormone (hGH) in serum plasma cell culture supernatants and other biological sample types This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate pre-coated with capture antibody detection antibody calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay kit offers a simple rapid and robust method to measure hGH levels