Learn More
Meso Scale Discovery RPLEX HUM HAVCR1/KIM1 ASYQP
SDP
Catalog No. 5000080288
Encompass
The R-PLEX Human HAVCR1 (KIM-1) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human kidney injury molecule 1 (KIM-1) in serum plasma urine cell culture supernatants and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls needed to quantify KIM-1 levels Recommended applications include biomarker research disease progression monitoring and drug discovery and development
Catalog No. 50-000-80288 Supplier Meso Scale Discovery Supplier No. K1514UR22
The R-PLEX Human HAVCR1 (KIM-1) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human kidney injury molecule 1 (KIM-1) in serum plasma urine cell culture supernatants and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls needed to quantify KIM-1 levels Recommended applications include biomarker research disease progression monitoring and drug discovery and development