Learn More
Meso Scale Discovery RPLEX HUM HCC4CCL16 ASYQP
SDP
Catalog No. 5000079500
Encompass
The R-PLEX Human HCC-4 (CCL16) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring 4 human hepatocellular carcinoma biomarkers in serum plasma and cell culture supernatants It quantifies HGF OPN PRL and suPAR using electrochemiluminescence on proprietary plates The kit supports high-throughput sensitive and reproducible quantification for monitoring carcinoma progression and therapy response It includes a 96-well plate calibrators controls detection antibodies read buffer wash buffer and instructions
Catalog No. 50-000-79500 Supplier Meso Scale Discovery Supplier No. K151AATR22
The R-PLEX Human HCC-4 (CCL16) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring 4 human hepatocellular carcinoma biomarkers in serum plasma and cell culture supernatants It quantifies HGF OPN PRL and suPAR using electrochemiluminescence on proprietary plates The kit supports high-throughput sensitive and reproducible quantification for monitoring carcinoma progression and therapy response It includes a 96-well plate calibrators controls detection antibodies read buffer wash buffer and instructions